missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Galectin-7 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 3 publications
£157.00 - £415.00
Specifications
| Antigen | Galectin-7 |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18777177
![]() |
R&D Systems
AF1304-SP |
25 μg |
£157.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18053152
|
R&D Systems
AF1304 |
100 μg |
£415.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Galectin-7 Polyclonal specifically detects Galectin-7 in Mouse samples. It is validated for Western Blot, Immunohistochemistry.Specifications
| Galectin-7 | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| Polyclonal | |
| Goat | |
| Mouse | |
| Gal-7, GAL7, galectin 7, galectin-7, Galectin7, HKL-14, lectin, galactoside-binding, soluble, 7, LGALS7, LGALS7A, p53-induced gene 1 protein, PI7, PIG1, TP53I1 | |
| LGALS7 | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| LYOPH | |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| RUO | |
| AAK29385 | |
| 3963 | |
| E. coli-derived recombinant mouse Galectin-7 Ser2-Phe136 Accession # AAK29385 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects mouse Galectin-7 in direct ELISAs and Western blots. In direct ELISAs, approximately 40% cross-reactivity with recombinant human Galectin-7 is observed and less than 1% cross-reactivity with recombinant mouse (rm) Galectin-1, rmGalectin-3, recombinant human (rh) Galectin-8, and rhGalectin-2 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title