missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Galectin-7 Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 3 publications
Brand: R&D Systems AF1304
This item is not returnable.
View return policy
Description
Galectin-7 Polyclonal specifically detects Galectin-7 in Mouse samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Galectin-7 | |
| Polyclonal | |
| Unconjugated | |
| AAK29385 | |
| LGALS7 | |
| E. coli-derived recombinant mouse Galectin-7 Ser2-Phe136 Accession # AAK29385 | |
| 100 μg | |
| Primary | |
| Detects mouse Galectin-7 in direct ELISAs and Western blots. In direct ELISAs, approximately 40% cross-reactivity with recombinant human Galectin-7 is observed and less than 1% cross-reactivity with recombinant mouse (rm) Galectin-1, rmGalectin-3, recombinant human (rh) Galectin-8, and rhGalectin-2 is observed. | |
| Mouse | |
| IgG |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| Gal-7, GAL7, galectin 7, galectin-7, Galectin7, HKL-14, lectin, galactoside-binding, soluble, 7, LGALS7, LGALS7A, p53-induced gene 1 protein, PI7, PIG1, TP53I1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 3963 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction