missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Fc gamma RIII (CD16) Antibody, R&D Systems™

Goat Polyclonal Antibody
£167.00 - £501.00
Specifications
| Antigen | Fc gamma RIII (CD16) |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 0.1 ug/mL, Blockade of Receptor-ligand Interaction, Immunocytochemistry 5-15 ug/mL |
| Applications | Western Blot, Blockade of Receptor-ligand Interaction, Immunocytochemistry |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18799027
![]() |
R&D Systems
AF1960-SP |
25 μg |
£167.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18021792
![]() |
R&D Systems
AF1960 |
100 μg |
£501.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Fc gamma RIII (CD16) Polyclonal specifically detects Fc gamma RIII (CD16) in Mouse samples. It is validated for Western Blot, Blockade of Receptor-ligand Interaction, Immunocytochemistry.Specifications
| Fc gamma RIII (CD16) | |
| Western Blot 0.1 ug/mL, Blockade of Receptor-ligand Interaction, Immunocytochemistry 5-15 ug/mL | |
| Polyclonal | |
| Goat | |
| Mouse | |
| P08508 | |
| 2214 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Fc gamma RIII Ala31-Thr215 Accession # P08508 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects Fc gamma RIIIA/B (CD16) in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 5% cross-reactivity with recombinant human (rh) Fc gamma RIIIB and rhCD32 is observed. |
| LYOPH | |
| Western Blot, Blockade of Receptor-ligand Interaction, Immunocytochemistry | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. with No Preservative | |
| CD16, CD16A, Fc fragment of IgG receptor IIIa, FCG3, FCGR3, FCGRIII, FCR-10, FCRIII, FCRIIIA, IGFR3, IMD20 | |
| Fcgr3 | |
| IgG | |
| Affinity Purified | |
| Store the unopened product at -20°C to -70 °C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title