missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Fc gamma RIII (CD16) Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF1960-SP
This item is not returnable.
View return policy
Description
Fc gamma RIII (CD16) Polyclonal specifically detects Fc gamma RIII (CD16) in Mouse samples. It is validated for Western Blot, Blockade of Receptor-ligand Interaction, Immunocytochemistry.
Specifications
| Fc gamma RIII (CD16) | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. with No Preservative | |
| CD16, CD16A, Fc fragment of IgG receptor IIIa, FCG3, FCGR3, FCGRIII, FCR-10, FCRIII, FCRIIIA, IGFR3, IMD20 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 2214 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Store the unopened product at -20°C to -70 °C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Western Blot, Blockade of Receptor-ligand Interaction, Immunocytochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Blockade of Receptor-ligand Interaction, Immunocytochemistry 5-15 ug/mL | |
| P08508 | |
| Fcgr3 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Fc gamma RIII Ala31-Thr215 Accession # P08508 | |
| 25 μg | |
| Primary | |
| Detects Fc gamma RIIIA/B (CD16) in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 5% cross-reactivity with recombinant human (rh) Fc gamma RIIIB and rhCD32 is observed. | |
| Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction