missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse FAM3C Antibody, R&D Systems™

Goat Polyclonal Antibody
£146.00 - £415.00
Specifications
| Antigen | FAM3C |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 0.1 ug/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18784559
![]() |
R&D Systems
AF2868-SP |
25 μg |
£146.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18012222
|
R&D Systems
AF2868 |
100 μg |
£415.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FAM3C Polyclonal specifically detects FAM3C in Mouse samples. It is validated for Western Blot.Specifications
| FAM3C | |
| Western Blot 0.1 ug/mL | |
| Polyclonal | |
| Goat | |
| Mouse | |
| D6WSU176e, family with sequence similarity 3, member C, GS3876, ILEI, ILEIInterleukin-like EMT inducer, predicted osteoblast protein | |
| FAM3C | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Q91VU0 | |
| 10447 | |
| Mouse myeloma cell line NS0-derived recombinant mouse FAM3C Gln25-Asp227 Accession # Q91VU0 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects mouse FAM3C in direct ELISAs and Western blots. In Western blots, approximately 50% cross-reactivity with recombinant human FAM3C is observed and less than 5% cross-reactivity with recombinant mouse (rm) FAM3B and rmFAM3D is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title