missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse FAM3C Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF2868-SP
This item is not returnable.
View return policy
Description
FAM3C Polyclonal specifically detects FAM3C in Mouse samples. It is validated for Western Blot.
Specifications
| FAM3C | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| D6WSU176e, family with sequence similarity 3, member C, GS3876, ILEI, ILEIInterleukin-like EMT inducer, predicted osteoblast protein | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 10447 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| Q91VU0 | |
| FAM3C | |
| Mouse myeloma cell line NS0-derived recombinant mouse FAM3C Gln25-Asp227 Accession # Q91VU0 | |
| 25 μg | |
| Primary | |
| Detects mouse FAM3C in direct ELISAs and Western blots. In Western blots, approximately 50% cross-reactivity with recombinant human FAM3C is observed and less than 5% cross-reactivity with recombinant mouse (rm) FAM3B and rmFAM3D is observed. | |
| Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction