missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Ephrin-A4 Antibody, R&D Systems™

Rat Monoclonal Antibody
£167.00 - £478.00
Specifications
| Antigen | Ephrin-A4 |
|---|---|
| Clone | 132416 |
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18749688
![]() |
R&D Systems
MAB569-SP |
25 μg |
£167.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18141051
![]() |
R&D Systems
MAB569 |
500 μg |
£478.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Ephrin-A4 Monoclonal specifically detects Ephrin-A4 in Mouse samples. It is validated for Western Blot.Specifications
| Ephrin-A4 | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EFL4, EFL-4, EFNA4, EPH-related receptor tyrosine kinase ligand 4, ephrin-A4, EphrinA4, EPLG4MGC125826, LERK-4, LERK4FLJ57652, ligand of eph-related kinase 4 | |
| EFNA4 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 132416 | |
| Western Blot | |
| Unconjugated | |
| Rat | |
| Mouse | |
| O08542 | |
| 1945 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse Ephrin-A4 Leu26-Gly176 Accession # O08542 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects mouse Ephrin-A4 in direct ELISAs and Western blots. In direct ELISAs, 100% cross-reactivity with recombinant human (rh) Ephrin-A4 is observed and no cross-reactivity with recombinant mouse (rm) Ephrin-A1, -A2, -B1, -B2, rhEphrin-A3, -A5, or -B3 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title