missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Ephrin-A4 Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB569
This item is not returnable.
View return policy
Description
Ephrin-A4 Monoclonal specifically detects Ephrin-A4 in Mouse samples. It is validated for Western Blot.
Specifications
| Ephrin-A4 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EFL4, EFL-4, EFNA4, EPH-related receptor tyrosine kinase ligand 4, ephrin-A4, EphrinA4, EPLG4MGC125826, LERK-4, LERK4FLJ57652, ligand of eph-related kinase 4 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 1945 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
| Western Blot | |
| 132416 | |
| Western Blot 1 ug/mL | |
| O08542 | |
| EFNA4 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse Ephrin-A4 Leu26-Gly176 Accession # O08542 | |
| 500 μg | |
| Primary | |
| Detects mouse Ephrin-A4 in direct ELISAs and Western blots. In direct ELISAs, 100% cross-reactivity with recombinant human (rh) Ephrin-A4 is observed and no cross-reactivity with recombinant mouse (rm) Ephrin-A1, -A2, -B1, -B2, rhEphrin-A3, -A5, or -B3 is observed. | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction