missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse EphA3 Antibody, R&D Systems™

Rat Monoclonal Antibody
£157.00 - £452.00
Specifications
| Antigen | EphA3 |
|---|---|
| Clone | 99437 |
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18776448
![]() |
R&D Systems
MAB640-SP |
25 μg |
£157.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18111201
|
R&D Systems
MAB640 |
500 μg |
£452.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
EphA3 Monoclonal specifically detects EphA3 in Mouse samples. It is validated for Western Blot.Specifications
| EphA3 | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Cek4, EC 2.7.10, EC 2.7.10.1, EK4, EPH receptor A3, EphA3, EPH-like kinase 4, ephrin type-A receptor 3, ETK, hEK4, HEKETK1eph-like tyrosine kinase 1, human embryo kinase 1, Tyro4, TYRO4 protein tyrosine kinase, Tyrosine-protein kinase receptor ETK1, Tyrosine-protein kinase TYRO4 | |
| EPHA3 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 99437 | |
| Western Blot | |
| Unconjugated | |
| Rat | |
| Mouse | |
| P29319 | |
| 2042 | |
| Mouse myeloma cell line NS0-derived recombinant mouse EphA3 Glu21-Val541 (predicted) Accession # P29319 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects mouse EphA3 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human (rh) EphA1, recombinant mouse (rm) EphA2, A4, A6, A7,A8, recombinant rat (rr) EphA5, rrEphB1, rmEphB2, B3, or B6 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title