missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse EphA3 Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB640-SP
This item is not returnable.
View return policy
Description
EphA3 Monoclonal specifically detects EphA3 in Mouse samples. It is validated for Western Blot.
Specifications
| EphA3 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Cek4, EC 2.7.10, EC 2.7.10.1, EK4, EPH receptor A3, EphA3, EPH-like kinase 4, ephrin type-A receptor 3, ETK, hEK4, HEKETK1eph-like tyrosine kinase 1, human embryo kinase 1, Tyro4, TYRO4 protein tyrosine kinase, Tyrosine-protein kinase receptor ETK1, Tyrosine-protein kinase TYRO4 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 2042 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
| Western Blot | |
| 99437 | |
| Western Blot 1 ug/mL | |
| P29319 | |
| EPHA3 | |
| Mouse myeloma cell line NS0-derived recombinant mouse EphA3 Glu21-Val541 (predicted) Accession # P29319 | |
| 25 μg | |
| Primary | |
| Detects mouse EphA3 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human (rh) EphA1, recombinant mouse (rm) EphA2, A4, A6, A7,A8, recombinant rat (rr) EphA5, rrEphB1, rmEphB2, B3, or B6 is observed. | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction