missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse EMMPRIN/CD147 Antibody, R&D Systems™

Rat Monoclonal Antibody
£167.00 - £487.00
Specifications
| Antigen | EMMPRIN/CD147 |
|---|---|
| Clone | 116318 |
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18712989
![]() |
R&D Systems
MAB772-SP |
25 μg |
£167.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18111531
![]() |
R&D Systems
MAB772 |
500 μg |
£487.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
EMMPRIN/CD147 Monoclonal specifically detects EMMPRIN/CD147 in Mouse samples. It is validated for Western Blot.Specifications
| EMMPRIN/CD147 | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 5F7, basigin, basigin (Ok blood group), BSG, CD147, CD147 antigen, Collagenase stimulatory factor, EMMPRINTCSF, Extracellular matrix metalloproteinase inducer, Leukocyte activation antigen M6, M6, OK, OK blood group antigen, Tumor cell-derived collagenase stimulatory factor | |
| BSG | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 116318 | |
| Western Blot | |
| Unconjugated | |
| Rat | |
| Mouse | |
| NP_001070652 | |
| 682 | |
| Mouse myeloma cell line NS0-derived recombinant mouse EMMPRIN/CD147 Ala22-Arg209 Accession # NP_001070652 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects mouse EMMPRIN/CD147 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human EMMPRIN is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title