missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse EMMPRIN/CD147 Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB772-SP
This item is not returnable.
View return policy
Description
EMMPRIN/CD147 Monoclonal specifically detects EMMPRIN/CD147 in Mouse samples. It is validated for Western Blot.
Specifications
| EMMPRIN/CD147 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 5F7, basigin, basigin (Ok blood group), BSG, CD147, CD147 antigen, Collagenase stimulatory factor, EMMPRINTCSF, Extracellular matrix metalloproteinase inducer, Leukocyte activation antigen M6, M6, OK, OK blood group antigen, Tumor cell-derived collagenase stimulatory factor | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 682 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
| Western Blot | |
| 116318 | |
| Western Blot 1 ug/mL | |
| NP_001070652 | |
| BSG | |
| Mouse myeloma cell line NS0-derived recombinant mouse EMMPRIN/CD147 Ala22-Arg209 Accession # NP_001070652 | |
| 25 μg | |
| Primary | |
| Detects mouse EMMPRIN/CD147 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human EMMPRIN is observed. | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction