missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse EDAR Antibody, R&D Systems™
Rat Monoclonal Antibody
£442.75
Specifications
Antigen | EDAR |
---|---|
Clone | 132106 |
Dilution | ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
Applications | ELISA |
Classification | Monoclonal |
Description
EDAR Monoclonal specifically detects EDAR in Mouse samples. It is validated for ELISA Capture (Matched Antibody Pair).Specifications
EDAR | |
ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Anhidrotic ectodysplasin receptor 1, DLED3, Ectodermal dysplasia receptor, ectodysplasin 1, anhidrotic receptor, ectodysplasin A receptor, Ectodysplasin-A receptor, ED1R, ED5, EDA1R, EDA3EDA-A1 receptor, EDA-A1R, Edar, FLJ94390, HRM1, mouse, homolog of, tumor necrosis factor receptor superfamily member EDAR | |
EDAR | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
132106 | |
ELISA | |
Unconjugated | |
Rat | |
Mouse | |
Q9R187 | |
10913 | |
Mouse myeloma cell line NS0-derived recombinant mouse EDAR Glu27-Ile189 Accession # Q9R187 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse EDAR in ELISAs. In sandwich immunoassays, less than 10% cross-reactivity with recombinant human EDAR is observed and no cross-reactivity with recombinant mouse (rm) EDA, rmTROY, or rhXEDAR is observed. |
For Research Use Only