missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse EDAR Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB7451
This item is not returnable.
View return policy
Description
EDAR Monoclonal specifically detects EDAR in Mouse samples. It is validated for ELISA Capture (Matched Antibody Pair).
Specifications
| EDAR | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Anhidrotic ectodysplasin receptor 1, DLED3, Ectodermal dysplasia receptor, ectodysplasin 1, anhidrotic receptor, ectodysplasin A receptor, Ectodysplasin-A receptor, ED1R, ED5, EDA1R, EDA3EDA-A1 receptor, EDA-A1R, Edar, FLJ94390, HRM1, mouse, homolog of, tumor necrosis factor receptor superfamily member EDAR | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 10913 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
| ELISA | |
| 132106 | |
| ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Q9R187 | |
| EDAR | |
| Mouse myeloma cell line NS0-derived recombinant mouse EDAR Glu27-Ile189 Accession # Q9R187 | |
| 500 μg | |
| Primary | |
| Detects mouse EDAR in ELISAs. In sandwich immunoassays, less than 10% cross-reactivity with recombinant human EDAR is observed and no cross-reactivity with recombinant mouse (rm) EDA, rmTROY, or rhXEDAR is observed. | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction