missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Desert Hedgehog/Dhh N-Terminus Antibody, R&D Systems™

Rat Monoclonal Antibody
£167.00 - £496.00
Specifications
| Antigen | Desert Hedgehog/Dhh |
|---|---|
| Clone | 126925 |
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18793129
![]() |
R&D Systems
MAB733-SP |
25 μg |
£167.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18031972
![]() |
R&D Systems
MAB733 |
500 μg |
£496.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Desert Hedgehog/Dhh Monoclonal specifically detects Desert Hedgehog/Dhh in Mouse samples. It is validated for Western Blot.Specifications
| Desert Hedgehog/Dhh | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| desert hedgehog, desert hedgehog (Drosophila) homolog, desert hedgehog homolog, desert hedgehog protein, DHH, HHG-3MGC35145 | |
| DHH | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 126925 | |
| Western Blot | |
| Unconjugated | |
| Rat | |
| Mouse | |
| Q61488 | |
| 50846 | |
| E. coli-derived recombinant mouse Desert Hedgehog/Dhh Cys23-Gly396 Accession # Q61488 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects mouse Desert Hedgehog/Dhh N-Terminus in direct ELISAs and Western blots. In Western blots, no cross-reactivity with the N-terminal peptides of recombinant human Shh or recombinant mouse (rm) Shh is observed. Also, in direct ELISAs and Western blots, no cross-reactivity with the C-terminal peptides of rmDhh, rmIhh, or rmShh is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title