missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Desert Hedgehog/Dhh N-Terminus Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB733-SP
This item is not returnable.
View return policy
Description
Desert Hedgehog/Dhh Monoclonal specifically detects Desert Hedgehog/Dhh in Mouse samples. It is validated for Western Blot.
Specifications
| Desert Hedgehog/Dhh | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| desert hedgehog, desert hedgehog (Drosophila) homolog, desert hedgehog homolog, desert hedgehog protein, DHH, HHG-3MGC35145 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 50846 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
| Western Blot | |
| 126925 | |
| Western Blot 1 ug/mL | |
| Q61488 | |
| DHH | |
| E. coli-derived recombinant mouse Desert Hedgehog/Dhh Cys23-Gly396 Accession # Q61488 | |
| 25 μg | |
| Primary | |
| Detects mouse Desert Hedgehog/Dhh N-Terminus in direct ELISAs and Western blots. In Western blots, no cross-reactivity with the N-terminal peptides of recombinant human Shh or recombinant mouse (rm) Shh is observed. Also, in direct ELISAs and Western blots, no cross-reactivity with the C-terminal peptides of rmDhh, rmIhh, or rmShh is observed. | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction