missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse DAN Antibody, R&D Systems™

Rat Monoclonal Antibody
£167.00 - £487.00
Specifications
| Antigen | DAN/NBL1 |
|---|---|
| Clone | 123202 |
| Dilution | Western Blot 1 ug/mL, Blockade of Receptor-ligand Interaction |
| Applications | Western Blot, Blockade of Receptor-ligand Interaction |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18757029
![]() |
R&D Systems
MAB7551-SP |
25 μg |
£167.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18131481
![]() |
R&D Systems
MAB7551 |
500 μg |
£487.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DAN/NBL1 Monoclonal specifically detects DAN/NBL1 in Mouse samples. It is validated for Western Blot, Blockade of Receptor-ligand Interaction.Specifications
| DAN/NBL1 | |
| Western Blot 1 ug/mL, Blockade of Receptor-ligand Interaction | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| DAN domain family member 1, DAND1, MGC8972, NBL1, neuroblastoma suppressor of tumorigenicity 1, neuroblastoma, suppression of tumorigenicity 1, Protein N03, suppression of tumorigenicity 1, Zinc finger protein DAN | |
| NBL1 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 123202 | |
| Western Blot, Blockade of Receptor-ligand Interaction | |
| Unconjugated | |
| Rat | |
| Mouse | |
| Q61477.2 | |
| 4681 | |
| Mouse myeloma cell line NS0-derived recombinant mouse DAN Ala17-Asp178 Accession # Q61477.2 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects mouse DAN in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) DAN, recombinant chicken Caronte, rhCerberus, or recombinant mouse Gremlin is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title