missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse DAN Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB7551-SP
This item is not returnable.
View return policy
Description
DAN/NBL1 Monoclonal specifically detects DAN/NBL1 in Mouse samples. It is validated for Western Blot, Blockade of Receptor-ligand Interaction.
Specifications
| DAN/NBL1 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| DAN domain family member 1, DAND1, MGC8972, NBL1, neuroblastoma suppressor of tumorigenicity 1, neuroblastoma, suppression of tumorigenicity 1, Protein N03, suppression of tumorigenicity 1, Zinc finger protein DAN | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 4681 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
| Western Blot, Blockade of Receptor-ligand Interaction | |
| 123202 | |
| Western Blot 1 ug/mL, Blockade of Receptor-ligand Interaction | |
| Q61477.2 | |
| NBL1 | |
| Mouse myeloma cell line NS0-derived recombinant mouse DAN Ala17-Asp178 Accession # Q61477.2 | |
| 25 μg | |
| Primary | |
| Detects mouse DAN in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) DAN, recombinant chicken Caronte, rhCerberus, or recombinant mouse Gremlin is observed. | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction