missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CXCL15/Lungkine Antibody, R&D Systems™

Goat Polyclonal Antibody
£168.00 - £438.00
Specifications
| Antigen | CXCL15/Lungkine |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 0.1 ug/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18775668
![]() |
R&D Systems
AF442-SP |
25 μg |
£168.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18090112
|
R&D Systems
AF442 |
100 μg |
£438.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CXCL15/Lungkine Polyclonal specifically detects CXCL15/Lungkine in Mouse samples. It is validated for Western Blot.Specifications
| CXCL15/Lungkine | |
| Western Blot 0.1 ug/mL | |
| Polyclonal | |
| Goat | |
| Mouse | |
| lungkine, Scyb15, weche | |
| CXCL15 | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Q9WVL7 | |
| 20309 | |
| E. coli-derived recombinant mouse CXCL15/Lungkine (R&D Systems, Catalog # 442-LK) Gln26-Ala167 Accession # Q9WVL7 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects mouse CXCL15/Lungkine in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with rm6Ckine, rmC10, rrCINC-1, rmBLC, rmCRG-2, rmEotaxin, rmFractalkine, rmGCP-2, rmJE, rmKC, rmLymphotactin, rmMARC, rmMDC, rmMIG, rmMIP-1 alpha , rmMIP-1 beta , rmRANTES, rmSDF-1 alpha , rmTCA-3 and rmTeck is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title