missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CXCL15/Lungkine Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems AF442
This item is not returnable.
View return policy
Description
CXCL15/Lungkine Polyclonal specifically detects CXCL15/Lungkine in Mouse samples. It is validated for Western Blot.
Specifications
| CXCL15/Lungkine | |
| Polyclonal | |
| Unconjugated | |
| Q9WVL7 | |
| CXCL15 | |
| E. coli-derived recombinant mouse CXCL15/Lungkine (R&D Systems, Catalog # 442-LK) Gln26-Ala167 Accession # Q9WVL7 | |
| 100 μg | |
| Primary | |
| Detects mouse CXCL15/Lungkine in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with rm6Ckine, rmC10, rrCINC-1, rmBLC, rmCRG-2, rmEotaxin, rmFractalkine, rmGCP-2, rmJE, rmKC, rmLymphotactin, rmMARC, rmMDC, rmMIG, rmMIP-1 alpha , rmMIP-1 beta , rmRANTES, rmSDF-1 alpha , rmTCA-3 and rmTeck is observed. | |
| Mouse | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| lungkine, Scyb15, weche | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 20309 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction