missing translation for 'onlineSavingsMsg'
Learn More

Mouse CXCL13/BLC/BCA-1 Antibody, R&D Systems™
GREENER_CHOICE

Rat Monoclonal Antibody has been used in 2 publications

£168.00 - £1660.00

Specifications

Antigen CXCL13/BLC/BCA-1
Clone 143608
Dilution Western Blot 1 ug/mL, Neutralization 12-30 ug/mL
Applications Western Blot, Neutralization
Classification Monoclonal
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18751478
GREENER_CHOICE
View Documents
R&D Systems
MAB4701-SP
25 μg
£168.00
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18737509
GREENER_CHOICE
View Documents
R&D Systems
MAB4701-100
100 μg
£412.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18703378
GREENER_CHOICE
View Documents
R&D Systems
MAB4701-500
500 μg
£1660.00
500µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CXCL13/BLC/BCA-1 Monoclonal specifically detects CXCL13/BLC/BCA-1 in Mouse samples. It is validated for Western Blot, Neutralization.
Specifications

Specifications

CXCL13/BLC/BCA-1
Western Blot 1 ug/mL, Neutralization 12-30 ug/mL
Monoclonal
Purified
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
ANGIE, ANGIE2, B cell-attracting chemokine 1, B lymphocyte chemoattractant, BCA1, BCA-1, BCA1B-cell chemoattractant, BCA-1CXC chemokine BLC, B-cell-homing chemokine (ligand for Burkitt's lymphoma receptor-1), BLC, BLCSmall-inducible cytokine B13, BLR1L, B-lymphocyte chemoattractant, chemokine (C-X-C motif) ligand 13, chemokine (C-X-C motif) ligand 13 (B-cell chemoattractant), C-X-C motif chemokine 13, SCYB13, SCYB13B-cell-attracting chemokine 1, small inducible cytokine B subfamily (Cys-X-Cys motif), member 13 (B-cellchemoattractant)
CXCL13
IgG2a
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
143608
Western Blot, Neutralization
Unconjugated
Rat
Mouse
O55038
10563
E. coli-derived recombinant mouse CXCL13/BLC/BCA-1 Ile22-Ala109 Accession # O55038
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects mouse CXCL13/BLC/BCA-1 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant cotton rat CXCL10, recombinant human CXCL1, 2, 3, 4, 5, 6, 7, 8, 9, 10, 11, 12/SDF-1 alpha , 12/SDF-1 beta , 13, 16, recombinant mouse CXCL1, 2, 4, 6, 7, 9, 10, 11, 12/SDF-1 alpha , 15, 16, recombinant porcine CXCL8, recombinant rat CXCL1, 2, 3/CINC-2 alpha , rr3/CINC-2 beta , or 5 is observed.
Vidéos
FDS
Documents

Documents

Certificats

For Research Use Only

Correction du contenu d'un produit

Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.

Nom du produit

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.

Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis