missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CXCL13/BLC/BCA-1 Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 2 publications
Brand: R&D Systems MAB4701-100
This item is not returnable.
View return policy
Description
CXCL13/BLC/BCA-1 Monoclonal specifically detects CXCL13/BLC/BCA-1 in Mouse samples. It is validated for Western Blot, Neutralization.
Specifications
| CXCL13/BLC/BCA-1 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| ANGIE, ANGIE2, B cell-attracting chemokine 1, B lymphocyte chemoattractant, BCA1, BCA-1, BCA1B-cell chemoattractant, BCA-1CXC chemokine BLC, B-cell-homing chemokine (ligand for Burkitt's lymphoma receptor-1), BLC, BLCSmall-inducible cytokine B13, BLR1L, B-lymphocyte chemoattractant, chemokine (C-X-C motif) ligand 13, chemokine (C-X-C motif) ligand 13 (B-cell chemoattractant), C-X-C motif chemokine 13, SCYB13, SCYB13B-cell-attracting chemokine 1, small inducible cytokine B subfamily (Cys-X-Cys motif), member 13 (B-cellchemoattractant) | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 10563 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
| Western Blot, Neutralization | |
| 143608 | |
| Western Blot 1 ug/mL, Neutralization 12-30 ug/mL | |
| O55038 | |
| CXCL13 | |
| E. coli-derived recombinant mouse CXCL13/BLC/BCA-1 Ile22-Ala109 Accession # O55038 | |
| 100 μg | |
| Primary | |
| Detects mouse CXCL13/BLC/BCA-1 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant cotton rat CXCL10, recombinant human CXCL1, 2, 3, 4, 5, 6, 7, 8, 9, 10, 11, 12/SDF-1 alpha , 12/SDF-1 beta , 13, 16, recombinant mouse CXCL1, 2, 4, 6, 7, 9, 10, 11, 12/SDF-1 alpha , 15, 16, recombinant porcine CXCL8, recombinant rat CXCL1, 2, 3/CINC-2 alpha , rr3/CINC-2 beta , or 5 is observed. | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction