missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CXCL1/GRO alpha/KC/CINC-1 Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 46 publications
£187.00 - £1810.00
Specifications
| Antigen | CXCL1/GRO alpha/KC/CINC-1 |
|---|---|
| Clone | 48415 |
| Dilution | Western Blot 1 ug/mL, Neutralization 0.05-0.25 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
| Applications | Western Blot, Neutralization, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18744058
![]() |
R&D Systems
MAB453-SP |
25 μg |
£187.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18744999
![]() |
R&D Systems
MAB453-100 |
100 μg |
£438.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18777198
![]() |
R&D Systems
MAB453-500 |
500 μg |
£1810.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CXCL1/GRO alpha/KC/CINC-1 Monoclonal specifically detects CXCL1/GRO alpha/KC/CINC-1 in Mouse samples. It is validated for Western Blot, Neutralization, ELISA Capture (Matched Antibody Pair).Specifications
| CXCL1/GRO alpha/KC/CINC-1 | |
| Western Blot 1 ug/mL, Neutralization 0.05-0.25 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Monoclonal | |
| Rat | |
| Mouse | |
| P12850 | |
| 2919 | |
| E. coli-derived recombinant mouse CXCL1/GRO alpha/KC/CINC-1 Arg20-Lys96 Accession # P12850 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects mouse CXCL1/GRO alpha/KC/CINC-1 in direct ELISAs and Western blots. Shows 100% cross-reactivity with recombinant human (rh) CXCL2, less than 40% cross-reactivity with recombinant rat (rr) CINC-1, and no cross-reactivity with rrCINC-3, rhCXCL3, rhCXCL5, rhCXCL6, or recombinant mouse CXCL6. |
| 48415 | |
| Western Blot, Neutralization, ELISA | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CINC1, CINC-1, FSP, GRO alpha, GRO1, GROa, KC, MGSA, MGSA-a, MGSA-alpha, NAP-3, SCYB1 | |
| Cxcl1 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title