missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CXCL1/GRO alpha/KC/CINC-1 Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 46 publications
Brand: R&D Systems MAB453-500
Les retours ne sont pas autorisés pour ce produit.
Consulta la politica di reso
Descrizione
CXCL1/GRO alpha/KC/CINC-1 Monoclonal specifically detects CXCL1/GRO alpha/KC/CINC-1 in Mouse samples. It is validated for Western Blot, Neutralization, ELISA Capture (Matched Antibody Pair).
Specifica
| CXCL1/GRO alpha/KC/CINC-1 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CINC1, CINC-1, FSP, GRO alpha, GRO1, GROa, KC, MGSA, MGSA-a, MGSA-alpha, NAP-3, SCYB1 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 2919 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
| Western Blot, Neutralization, ELISA | |
| 48415 | |
| Western Blot 1 ug/mL, Neutralization 0.05-0.25 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| P12850 | |
| Cxcl1 | |
| E. coli-derived recombinant mouse CXCL1/GRO alpha/KC/CINC-1 Arg20-Lys96 Accession # P12850 | |
| 500 μg | |
| Primary | |
| Detects mouse CXCL1/GRO alpha/KC/CINC-1 in direct ELISAs and Western blots. Shows 100% cross-reactivity with recombinant human (rh) CXCL2, less than 40% cross-reactivity with recombinant rat (rr) CINC-1, and no cross-reactivity with rrCINC-3, rhCXCL3, rhCXCL5, rhCXCL6, or recombinant mouse CXCL6. | |
| Mouse | |
| Supernatant |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto