missing translation for 'onlineSavingsMsg'
Learn More

Mouse Cerberus 1 Antibody, R&D Systems™
GREENER_CHOICE

Rat Monoclonal Antibody has been used in 2 publications

£170.00 - £496.00

Specifications

Antigen Cerberus 1
Clone 225807
Concentration LYOPH
Dilution Western Blot 1 ug/mL
Applications Western Blot
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18722417
GREENER_CHOICE
View Documents
R&D Systems
MAB1986-SP
25 μg
£170.00
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18169950
View Documents
R&D Systems
MAB1986
500 μg
£496.00
500µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Cerberus 1 Monoclonal specifically detects Cerberus 1 in Mouse samples. It is validated for Western Blot.
Specifications

Specifications

Cerberus 1
LYOPH
Western Blot
Unconjugated
Rat
Mouse
O55233
9350
Mouse myeloma cell line NS0-derived recombinant mouse Cerberus 1 Asp18-Pro272 Accession # O55233
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects mouse Cerberus 1 in direct ELISAs and Western blots. In direct ELISAs, this antibody does not cross-react with recombinant human Cerberus, recombinant mouse (rm) DAN, or rmGremlin.
225807
Western Blot 1 ug/mL
Monoclonal
Purified
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
CER1, cerberus, cerberus 1 (Xenopus laevis) homolog (cysteine knot superfamily), cerberus 1, cysteine knot superfamily, homolog (Xenopus laevis), cerberus-related 1, Cerberus-related protein, DAN domain family member 4, DAND4, MGC119894, MGC119895, MGC96951
CER1
IgG2a
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.