missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Cerberus 1 Antibody, R&D Systems™
Rat Monoclonal Antibody has been used in 2 publications
Brand: R&D Systems MAB1986
This item is not returnable.
View return policy
Description
Cerberus 1 Monoclonal specifically detects Cerberus 1 in Mouse samples. It is validated for Western Blot.
Specifications
| Cerberus 1 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| O55233 | |
| CER1 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Cerberus 1 Asp18-Pro272 Accession # O55233 | |
| 500 μg | |
| Primary | |
| Detects mouse Cerberus 1 in direct ELISAs and Western blots. In direct ELISAs, this antibody does not cross-react with recombinant human Cerberus, recombinant mouse (rm) DAN, or rmGremlin. | |
| Mouse | |
| Purified |
| Western Blot | |
| 225807 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CER1, cerberus, cerberus 1 (Xenopus laevis) homolog (cysteine knot superfamily), cerberus 1, cysteine knot superfamily, homolog (Xenopus laevis), cerberus-related 1, Cerberus-related protein, DAN domain family member 4, DAND4, MGC119894, MGC119895, MGC96951 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 9350 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction