missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CD69 Antibody, R&D Systems™

Rat Monoclonal Antibody
£168.00 - £362.00
Specifications
| Antigen | CD69 |
|---|---|
| Clone | 310106 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells |
| Applications | Western Blot, Flow Cytometry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18710048
![]() |
R&D Systems
MAB2386-SP |
25 μg |
£168.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18129660
|
R&D Systems
MAB2386 |
100 μg |
£362.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD69 Monoclonal specifically detects CD69 in Mouse samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.Specifications
| CD69 | |
| LYOPH | |
| Western Blot, Flow Cytometry | |
| Unconjugated | |
| Rat | |
| Mouse | |
| P37217 | |
| 969 | |
| Mouse myeloma cell line NS0-derived recombinant mouse CD69 Gly64-Arg199 Accession # P37217 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects mouse CD69 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human CD69 is observed. |
| 310106 | |
| Western Blot 1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Activation inducer molecule, activation inducer molecule (AIM/CD69), AIM, BL-AC/P26, CD69, CD69 antigen, CD69 antigen (p60, early T-cell activation antigen), CD69 molecule, CLEC2CC-type lectin domain family 2 member C, C-type lectin domain family 2, member C, EA1, EA-1, early activation antigen CD69, early lymphocyte activation antigen, Early T-cell activation antigen p60, GP32/28, Leu23, Leukocyte surface antigen Leu-23, MLR-3, p60 | |
| CD69 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title