missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CD69 Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB2386-SP
This item is not returnable.
View return policy
Description
CD69 Monoclonal specifically detects CD69 in Mouse samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.
Specifications
| CD69 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells | |
| P37217 | |
| CD69 | |
| Mouse myeloma cell line NS0-derived recombinant mouse CD69 Gly64-Arg199 Accession # P37217 | |
| 25 μg | |
| Primary | |
| Detects mouse CD69 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human CD69 is observed. | |
| Mouse | |
| Purified |
| Western Blot, Flow Cytometry | |
| 310106 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Activation inducer molecule, activation inducer molecule (AIM/CD69), AIM, BL-AC/P26, CD69, CD69 antigen, CD69 antigen (p60, early T-cell activation antigen), CD69 molecule, CLEC2CC-type lectin domain family 2 member C, C-type lectin domain family 2, member C, EA1, EA-1, early activation antigen CD69, early lymphocyte activation antigen, Early T-cell activation antigen p60, GP32/28, Leu23, Leukocyte surface antigen Leu-23, MLR-3, p60 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 969 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction