missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CD3 epsilon Antibody, R&D Systems™

Hamster Monoclonal Antibody has been used in 14 publications
£178.00 - £1775.00
Specifications
| Antigen | CD3 epsilon |
|---|---|
| Clone | 145-2C11 |
| Dilution | Flow Cytometry 2.5 ug/10^6 cells, Immunoprecipitation, Cell depletion, T Cell Stimulation 0.1-0.6 ug/mL, CyTOF-reported |
| Applications | Flow Cytometry, Immunoprecipitation, Depletion Assay, T-Cell Activation, CyTOF |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18701708
![]() |
R&D Systems
MAB484-SP |
25 μg |
£178.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18766439
![]() |
R&D Systems
MAB484-100 |
100 μg |
£452.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18728049
![]() |
R&D Systems
MAB484-500 |
500 μg |
£1775.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD3 epsilon Monoclonal specifically detects CD3 epsilon in Mouse samples. It is validated for Flow Cytometry, Immunoprecipitation, Cell depletion, T Cell Stimulation, CyTOF-reported.Specifications
| CD3 epsilon | |
| Flow Cytometry 2.5 ug/10^6 cells, Immunoprecipitation, Cell depletion, T Cell Stimulation 0.1-0.6 ug/mL, CyTOF-reported | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 916 | |
| H-2K b -specific mouse cytotoxic T-lymphocyte | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects mouse CD3 epsilon . Binds to lymphocytes from all mouse strains tested and does not react with cells from rats, rabbits, miniature swine, or hamsters. 1 Binds to the CD3 epsilon-chain present on T-lymphocyes and thymocytes. Its binding has been characterized with respect to several other monoclonal anti-CD3 antibodies. 3,4 |
| 145-2C11 | |
| Flow Cytometry, Immunoprecipitation, Depletion Assay, T-Cell Activation, CyTOF | |
| Unconjugated | |
| Hamster | |
| Mouse | |
| CD3e, CD3e antigen, CD3e antigen, epsilon polypeptide (TiT3 complex), CD3e molecule, epsilon (CD3-TCR complex), CD3-epsilon, FLJ18683, T3E, T-cell antigen receptor complex, epsilon subunit of T3, T-cell surface antigen T3/Leu-4 epsilon chain, T-cell surface glycoprotein CD3 epsilon chain, TCRE | |
| Cd247 | |
| IgG | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title