missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CD3 epsilon Antibody, R&D Systems™

Hamster Monoclonal Antibody has been used in 14 publications
Brand: R&D Systems MAB484-SP
This item is not returnable.
View return policy
Description
CD3 epsilon Monoclonal specifically detects CD3 epsilon in Mouse samples. It is validated for Flow Cytometry, Immunoprecipitation, Cell depletion, T Cell Stimulation, CyTOF-reported.
Specifications
| CD3 epsilon | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Cd247 | |
| H-2K b -specific mouse cytotoxic T-lymphocyte | |
| 25 μg | |
| Primary | |
| Detects mouse CD3 epsilon . Binds to lymphocytes from all mouse strains tested and does not react with cells from rats, rabbits, miniature swine, or hamsters. 1 Binds to the CD3 epsilon-chain present on T-lymphocyes and thymocytes. Its binding has been characterized with respect to several other monoclonal anti-CD3 antibodies. 3,4 | |
| Mouse | |
| Purified |
| Flow Cytometry, Immunoprecipitation, Depletion Assay, T-Cell Activation, CyTOF | |
| 145-2C11 | |
| Flow Cytometry 2.5 ug/10^6 cells, Immunoprecipitation, Cell depletion, T Cell Stimulation 0.1-0.6 ug/mL, CyTOF-reported | |
| CD3e, CD3e antigen, CD3e antigen, epsilon polypeptide (TiT3 complex), CD3e molecule, epsilon (CD3-TCR complex), CD3-epsilon, FLJ18683, T3E, T-cell antigen receptor complex, epsilon subunit of T3, T-cell surface antigen T3/Leu-4 epsilon chain, T-cell surface glycoprotein CD3 epsilon chain, TCRE | |
| Hamster | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 916 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction