missing translation for 'onlineSavingsMsg'
Learn More

Mouse CD11b/Integrin alpha M Antibody, R&D Systems™
GREENER_CHOICE

Rabbit Monoclonal Antibody

£181.00 - £452.00

Specifications

Antigen CD11b
Clone 1250D
Dilution Western Blot 0.5 ug/mL, Simple Western 5 ug/mL
Applications Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18792697
GREENER_CHOICE
View Documents
R&D Systems
MAB11242-SP
25 μg
£181.00
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18761457
GREENER_CHOICE
View Documents
R&D Systems
MAB11242-100
100 μg
£452.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CD11b Monoclonal antibody specifically detects CD11b in Mouse samples. It is validated for Western Blot, Western Blot
Specifications

Specifications

CD11b
Western Blot 0.5 ug/mL, Simple Western 5 ug/mL
Monoclonal
Purified
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
antigen CD11b (p170), CD11 antigen-like family member B, CD11b, CD11b antigen, Cell surface glycoprotein MAC-1 subunit alpha, Complement component 3 receptor 3 subunit, CR-3 alpha chain, CR3A, CR3AMGC117044, Integrin alpha M, integrin alpha-M, integrin, alpha M (complement component 3 receptor 3 subunit), ITGAM, Leukocyte adhesion receptor MO1, MAC-1, MAC1A, macrophage antigen alpha polypeptide, MO1A, Neutrophil adherence receptor, neutrophil adherence receptor alpha-M subunit, SLEB6
MouseCD11b/Integrin alpha M synthetic peptide, Accession # P11215
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
1250D
Western Blot
Unconjugated
Rabbit
Mouse
P11215
3684
IgG
Protein A or G purified from cell culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Vidéos
FDS
Documents

Documents

Certificats
Correction du contenu d'un produit

Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.

Nom du produit

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.

Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis