missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CD11b/Integrin alpha M Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Brand: R&D Systems MAB11242-100
This item is not returnable.
View return policy
Description
CD11b Monoclonal antibody specifically detects CD11b in Mouse samples. It is validated for Western Blot, Western Blot
Specifications
| CD11b | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| antigen CD11b (p170), CD11 antigen-like family member B, CD11b, CD11b antigen, Cell surface glycoprotein MAC-1 subunit alpha, Complement component 3 receptor 3 subunit, CR-3 alpha chain, CR3A, CR3AMGC117044, Integrin alpha M, integrin alpha-M, integrin, alpha M (complement component 3 receptor 3 subunit), ITGAM, Leukocyte adhesion receptor MO1, MAC-1, MAC1A, macrophage antigen alpha polypeptide, MO1A, Neutrophil adherence receptor, neutrophil adherence receptor alpha-M subunit, SLEB6 | |
| MouseCD11b/Integrin alpha M synthetic peptide, Accession # P11215 | |
| 100 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
| IgG |
| Western Blot | |
| 1250D | |
| Western Blot 0.5 ug/mL, Simple Western 5 ug/mL | |
| P11215 | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 3684 | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction