missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CCL28 Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 7 publications
£168.00 - £1775.00
Specifications
| Antigen | CCL28 |
|---|---|
| Clone | 134306 |
| Dilution | Western Blot 1 ug/mL, Neutralization 2.5-5.0 ug/mL |
| Applications | Western Blot, Neutralization |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18791958
![]() |
R&D Systems
MAB533-SP |
25 μg |
£168.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18765519
![]() |
R&D Systems
MAB533-100 |
100 μg |
£454.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18748399
![]() |
R&D Systems
MAB533-500 |
500 μg |
£1775.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CCL28 Monoclonal specifically detects CCL28 in Mouse samples. It is validated for Western Blot, Neutralization.Specifications
| CCL28 | |
| Western Blot 1 ug/mL, Neutralization 2.5-5.0 ug/mL | |
| Monoclonal | |
| Rat | |
| Mouse | |
| Q9JIL2 | |
| 56477 | |
| E. coli-derived recombinant mouse CCL28 Ile23-Arg130 Accession # Q9JIL2 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects mouse CCL28 in Western blots. In Western blots, approximately 25% cross-reactivity with recombinant human (rh) CCL28 is observed and no cross-reactivity with rhCCL1, 2, 3, 4, 5, 7, 8, 11, 13, 14, 15, 16, 17, 18, 19, 20, 21, 22, 23, 24, 25, recombinant mouse CCL1, 2, 3, 4, 6, 7, 9/10, 11, 12, 19, 20, 21, 22, 24, 25, 27, or recombinant rat CCL20 is observed. |
| 134306 | |
| Western Blot, Neutralization | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CCL28, chemokine (C-C motif) ligand 28, MEC, member 28, MGC71902, Protein CCK1, VIC | |
| CCL28 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title