missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CCL28 Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 7 publications
Brand: R&D Systems MAB533-SP
This item is not returnable.
View return policy
Description
CCL28 Monoclonal specifically detects CCL28 in Mouse samples. It is validated for Western Blot, Neutralization.
Specifications
| CCL28 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CCL28, chemokine (C-C motif) ligand 28, MEC, member 28, MGC71902, Protein CCK1, VIC | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 56477 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
| Western Blot, Neutralization | |
| 134306 | |
| Western Blot 1 ug/mL, Neutralization 2.5-5.0 ug/mL | |
| Q9JIL2 | |
| CCL28 | |
| E. coli-derived recombinant mouse CCL28 Ile23-Arg130 Accession # Q9JIL2 | |
| 25 μg | |
| Primary | |
| Detects mouse CCL28 in Western blots. In Western blots, approximately 25% cross-reactivity with recombinant human (rh) CCL28 is observed and no cross-reactivity with rhCCL1, 2, 3, 4, 5, 7, 8, 11, 13, 14, 15, 16, 17, 18, 19, 20, 21, 22, 23, 24, 25, recombinant mouse CCL1, 2, 3, 4, 6, 7, 9/10, 11, 12, 19, 20, 21, 22, 24, 25, 27, or recombinant rat CCL20 is observed. | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction