missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Cathepsin F Antibody, R&D Systems™

Goat Polyclonal Antibody
£168.00 - £432.00
Specifications
| Antigen | Cathepsin F |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Immunoprecipitation 25 ug/mL |
| Applications | Western Blot, Immunohistochemistry, Immunoprecipitation |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18738097
![]() |
R&D Systems
AF2544-SP |
25 μg |
£168.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18001962
|
R&D Systems
AF2544 |
100 μg |
£432.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cathepsin F Polyclonal specifically detects Cathepsin F in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation.Specifications
| Cathepsin F | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Immunoprecipitation 25 ug/mL | |
| Polyclonal | |
| Goat | |
| Mouse | |
| cathepsin F, CATSFEC 3.4.22.41, CTSF, EC 3.4.22 | |
| CTSF | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| LYOPH | |
| Western Blot, Immunohistochemistry, Immunoprecipitation | |
| Unconjugated | |
| RUO | |
| Q9R013 | |
| 8722 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Cathepsin F Ser20-Asn462 Accession # Q9R013 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects mouse Cathepsin F in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant mouse (rm) Cathepsin C, rmCathepsin H, and rmCathepsin L is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title