missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Cathepsin F Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF2544-SP
This item is not returnable.
View return policy
Description
Cathepsin F Polyclonal specifically detects Cathepsin F in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation.
Specifications
| Cathepsin F | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| cathepsin F, CATSFEC 3.4.22.41, CTSF, EC 3.4.22 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 8722 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunohistochemistry, Immunoprecipitation | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Immunoprecipitation 25 ug/mL | |
| Q9R013 | |
| CTSF | |
| Mouse myeloma cell line NS0-derived recombinant mouse Cathepsin F Ser20-Asn462 Accession # Q9R013 | |
| 25 μg | |
| Primary | |
| Detects mouse Cathepsin F in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant mouse (rm) Cathepsin C, rmCathepsin H, and rmCathepsin L is observed. | |
| Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction