missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Caspase-11/Caspase-4 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
£178.00 - £379.00
Specifications
| Antigen | Caspase-11/Caspase-4 |
|---|---|
| Clone | 1172C |
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18772438
![]() |
R&D Systems
MAB8648-SP |
25 μg |
£178.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18702859
![]() |
R&D Systems
MAB8648 |
100 μg |
£379.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Caspase-11/Caspase-4 Monoclonal specifically detects Caspase-11/Caspase-4 in Mouse samples. It is validated for Western Blot.Specifications
| Caspase-11/Caspase-4 | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with Sodium Azide | |
| 12363 | |
| HEK293 human embryonic kidney cell line transfected with mouse Caspase-11/Caspase-4 Accession # P70343 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects mouse Caspase-11/Caspase-4 in Western blots. |
| 1172C | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Mouse | |
| P70343 | |
| Casp4 | |
| IgG | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title