missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Caspase-11/Caspase-4 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Brand: R&D Systems MAB8648-SP
This item is not returnable.
View return policy
Description
Caspase-11/Caspase-4 Monoclonal specifically detects Caspase-11/Caspase-4 in Mouse samples. It is validated for Western Blot.
Specifications
| Caspase-11/Caspase-4 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with Sodium Azide | |
| Casp4 | |
| HEK293 human embryonic kidney cell line transfected with mouse Caspase-11/Caspase-4 Accession # P70343 | |
| 25 μg | |
| Primary | |
| Detects mouse Caspase-11/Caspase-4 in Western blots. | |
| Mouse | |
| Purified |
| Western Blot | |
| 1172C | |
| Western Blot 1 ug/mL | |
| P70343 | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 12363 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction