missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse BTN2A2/Butyrophilin 2A2 Antibody, R&D Systems™

Sheep Polyclonal Antibody
£168.00 - £451.00
Specifications
| Antigen | BTN2A2/Butyrophilin 2 |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 0.1 ug/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18785837
![]() |
R&D Systems
AF4917-SP |
25 μg |
£168.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18000522
|
R&D Systems
AF4917 |
100 μg |
£451.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
BTN2A2/Butyrophilin 2 Polyclonal specifically detects BTN2A2/Butyrophilin 2 in Mouse samples. It is validated for Western Blot.Specifications
| BTN2A2/Butyrophilin 2 | |
| Western Blot 0.1 ug/mL | |
| Polyclonal | |
| Sheep | |
| Mouse | |
| BTF2, BTF2BT2.2butyrophilin 2, BTN2, Butyrophilin 2, butyrophilin subfamily 2 member A2, butyrophilin, subfamily 2, member A2, FLJ41908 | |
| BTN2A2 | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| A4QPC6.1 | |
| 10385 | |
| Mouse myeloma cell line NS0-derived recombinant mouse BTN2A2/Butyrophilin 2A2 Gln29-Ser219 Accession # A4QPC6.1 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects mouse BTN2A2/Butyrophilin 2A2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 10% cross-reactivity with recombinant mouse (rm) BTN and rmBTNL2 is observed. |
For Research Use Only
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto