missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse BTN2A2/Butyrophilin 2A2 Antibody, R&D Systems™
Sheep Polyclonal Antibody
Brand: R&D Systems AF4917
This item is not returnable.
View return policy
Description
BTN2A2/Butyrophilin 2 Polyclonal specifically detects BTN2A2/Butyrophilin 2 in Mouse samples. It is validated for Western Blot.
Specifications
| BTN2A2/Butyrophilin 2 | |
| Polyclonal | |
| Unconjugated | |
| A4QPC6.1 | |
| BTN2A2 | |
| Mouse myeloma cell line NS0-derived recombinant mouse BTN2A2/Butyrophilin 2A2 Gln29-Ser219 Accession # A4QPC6.1 | |
| 100 μg | |
| Primary | |
| Detects mouse BTN2A2/Butyrophilin 2A2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 10% cross-reactivity with recombinant mouse (rm) BTN and rmBTNL2 is observed. | |
| Mouse | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| BTF2, BTF2BT2.2butyrophilin 2, BTN2, Butyrophilin 2, butyrophilin subfamily 2 member A2, butyrophilin, subfamily 2, member A2, FLJ41908 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 10385 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction