missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse BLAME/SLAMF8 Antibody, R&D Systems™

Sheep Polyclonal Antibody
£168.00 - £451.00
Specifications
| Antigen | BLAME/SLAMF8 |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 0.1 ug/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18719107
![]() |
R&D Systems
AF4156-SP |
25 μg |
£168.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18020862
![]() |
R&D Systems
AF4156 |
100 μg |
£451.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
BLAME/SLAMF8 Polyclonal specifically detects BLAME/SLAMF8 in Mouse samples. It is validated for Western Blot.Specifications
| BLAME/SLAMF8 | |
| Western Blot 0.1 ug/mL | |
| Polyclonal | |
| Sheep | |
| Mouse | |
| B Lymphocyte Activator Macrophage Expressed, BCM-Like Membrane Protein, B-Lymphocyte Activator Macrophage Expressed, CD353, CD353 Antigen, SBBI42, SLAM Family Member 8, SLAMF8 | |
| Mouse myeloma cell line NS0-derived recombinant mouse BLAME/SLAMF8 Val21-Asp231 Accession # Q9D3G2 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects mouse BLAME/SLAMF8 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 40% cross-reactivity with recombinant human BLAME is observed. |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Q9D3G2 | |
| 56833 | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title