missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse BLAME/SLAMF8 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF4156
This item is not returnable.
View return policy
Description
BLAME/SLAMF8 Polyclonal specifically detects BLAME/SLAMF8 in Mouse samples. It is validated for Western Blot.
Specifications
| BLAME/SLAMF8 | |
| Polyclonal | |
| Unconjugated | |
| Q9D3G2 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 56833 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| B Lymphocyte Activator Macrophage Expressed, BCM-Like Membrane Protein, B-Lymphocyte Activator Macrophage Expressed, CD353, CD353 Antigen, SBBI42, SLAM Family Member 8, SLAMF8 | |
| Mouse myeloma cell line NS0-derived recombinant mouse BLAME/SLAMF8 Val21-Asp231 Accession # Q9D3G2 | |
| 100 μg | |
| Primary | |
| Detects mouse BLAME/SLAMF8 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 40% cross-reactivity with recombinant human BLAME is observed. | |
| Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction