missing translation for 'onlineSavingsMsg'
Learn More

Mouse Betacellulin/BTC Antibody, R&D Systems™
GREENER_CHOICE

Rat Monoclonal Antibody

£156.00 - £452.00

Specifications

Antigen Betacellulin/BTC
Clone 197417
Concentration LYOPH
Dilution Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Applications Western Blot, ELISA
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18739438
GREENER_CHOICE
View Documents
R&D Systems
MAB10251-SP
25 μg
£156.00
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18136081
View Documents
R&D Systems
MAB10251
500 μg
£452.00
500µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Betacellulin/BTC Monoclonal specifically detects Betacellulin/BTC in Mouse samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).
Specifications

Specifications

Betacellulin/BTC
LYOPH
Western Blot, ELISA
Unconjugated
Rat
Mouse
Q05928
685
E. coli-derived recombinant mouse Betacellulin Asp32-Gln118 Accession # Q05928
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects mouse Betacellulin in ELISAs and Western blots. In ELISAs and Western blots, this antibody does not cross-react with recombinant mouse (rm) Epigen, rhAR, rhBetacellulin, rhEGF, rhEpiregulin, rhNRG-2, rhNRG1-beta 1, or rhTGF-alpha .
197417
Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Monoclonal
Purified
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
betacellulin, BTC, probetacellulin
BTC
IgG2a
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.