missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Betacellulin/BTC Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB10251-SP
This item is not returnable.
View return policy
Description
Betacellulin/BTC Monoclonal specifically detects Betacellulin/BTC in Mouse samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).
Specifications
| Betacellulin/BTC | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Q05928 | |
| BTC | |
| E. coli-derived recombinant mouse Betacellulin Asp32-Gln118 Accession # Q05928 | |
| 25 μg | |
| Primary | |
| Detects mouse Betacellulin in ELISAs and Western blots. In ELISAs and Western blots, this antibody does not cross-react with recombinant mouse (rm) Epigen, rhAR, rhBetacellulin, rhEGF, rhEpiregulin, rhNRG-2, rhNRG1-beta 1, or rhTGF-alpha . | |
| Mouse | |
| Purified |
| Western Blot, ELISA | |
| 197417 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| betacellulin, BTC, probetacellulin | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 685 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction