missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse ASAH2/N-acylsphingosine Amidohydrolase-2 Antibody, R&D Systems™

Sheep Polyclonal Antibody
£168.00 - £417.00
Especificaciones
| Antigen | ASAH2/N-acylsphingosine Amidohydrolase-2 |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL |
| Applications | Western Blot, Immunoprecipitation |
| Classification | Polyclonal |
| Código de producto | Marca | Quantity | Precio | Cantidad y disponibilidad | |||||
|---|---|---|---|---|---|---|---|---|---|
| Código de producto | Marca | Quantity | Precio | Cantidad y disponibilidad | |||||
18714937
![]() |
R&D Systems
AF3558-SP |
25 μg |
£168.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18042032
|
R&D Systems
AF3558 |
100 μg |
£417.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Descripción
ASAH2/N-acylsphingosine Amidohydrolase-2 Polyclonal specifically detects ASAH2/N-acylsphingosine Amidohydrolase-2 in Mouse samples. It is validated for Western Blot, Immunoprecipitation.Especificaciones
| ASAH2/N-acylsphingosine Amidohydrolase-2 | |
| Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL | |
| Polyclonal | |
| Sheep | |
| Mouse | |
| ASAH2 N-acylsphingosine amidohydrolase (non-lysosomal ceramidase) 2, BCDase, HNAC1, LCDase, Nacylsphingosine Amidohydrolase2, NCDase, N-CDase | |
| ASAH2 | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| LYOPH | |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| RUO | |
| Q9JHE3 | |
| 56624 | |
| Mouse myeloma cell line NS0-derived recombinant mouse ASAH2/N-acylsphingosine Amidohydrolase-2 Thr34-Thr756 Accession # Q9JHE3 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects mouse ASAH2/N-acylsphingosine Amidohydrolase-2 in direct ELISAs and Western blots. In direct ELISAs, approximately 15% cross-reactivity with recombinant human ASAH2 is observed. |
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto