missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse ASAH2/N-acylsphingosine Amidohydrolase-2 Antibody, R&D Systems™
Sheep Polyclonal Antibody
Brand: R&D Systems AF3558
This item is not returnable.
View return policy
Description
ASAH2/N-acylsphingosine Amidohydrolase-2 Polyclonal specifically detects ASAH2/N-acylsphingosine Amidohydrolase-2 in Mouse samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| ASAH2/N-acylsphingosine Amidohydrolase-2 | |
| Polyclonal | |
| Unconjugated | |
| Q9JHE3 | |
| ASAH2 | |
| Mouse myeloma cell line NS0-derived recombinant mouse ASAH2/N-acylsphingosine Amidohydrolase-2 Thr34-Thr756 Accession # Q9JHE3 | |
| 100 μg | |
| Primary | |
| Detects mouse ASAH2/N-acylsphingosine Amidohydrolase-2 in direct ELISAs and Western blots. In direct ELISAs, approximately 15% cross-reactivity with recombinant human ASAH2 is observed. | |
| Mouse | |
| IgG |
| Western Blot, Immunoprecipitation | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL | |
| ASAH2 N-acylsphingosine amidohydrolase (non-lysosomal ceramidase) 2, BCDase, HNAC1, LCDase, Nacylsphingosine Amidohydrolase2, NCDase, N-CDase | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 56624 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction