missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Angiopoietin-3 Antibody, R&D Systems™

Rat Monoclonal Antibody
£146.00 - £451.00
Specifications
| Antigen | Angiopoietin-3 |
|---|---|
| Clone | 113538 |
| Concentration | LYOPH |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18765709
![]() |
R&D Systems
MAB1361-SP |
25 μg |
£146.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18120171
|
R&D Systems
MAB1361 |
500 μg |
£451.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Angiopoietin-3 Monoclonal specifically detects Angiopoietin-3 in Mouse samples. It is validated for .Specifications
| Angiopoietin-3 | |
| LYOPH | |
| Unconjugated | |
| Rat | |
| Mouse | |
| Q9WVH6 | |
| 51378 | |
| E. coli-derived recombinant mouse Angiopoietin-3 Ala21-Ala509 Accession # Q9WVH6 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects mouse Angiopoietin-3 in direct ELISAs and Western blots. |
| 113538 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| ANG3, ANG4, ANGPT3 | |
| Angpt4 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title