missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Angiopoietin-3 Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB1361-SP
This item is not returnable.
View return policy
Description
Angiopoietin-3 Monoclonal specifically detects Angiopoietin-3 in Mouse samples. It is validated for .
Specifications
| Angiopoietin-3 | |
| Monoclonal | |
| LYOPH | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| ANG3, ANG4, ANGPT3 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 51378 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
| ELISA | |
| 113538 | |
| Unconjugated | |
| Q9WVH6 | |
| Angpt4 | |
| E. coli-derived recombinant mouse Angiopoietin-3 Ala21-Ala509 Accession # Q9WVH6 | |
| 25 μg | |
| Primary | |
| Detects mouse Angiopoietin-3 in direct ELISAs and Western blots. | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction