missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse A33 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF2756
This item is not returnable.
View return policy
Description
A33/GPA33 Polyclonal specifically detects A33/GPA33 in Mouse samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| A33/GPA33 | |
| Polyclonal | |
| Unconjugated | |
| Q9JKA5 | |
| GPA33 | |
| Mouse myeloma cell line NS0-derived recombinant mouse A33 Leu22-Ile235 Accession # Q9JKA5 | |
| 100 μg | |
| Primary | |
| Detects mouse A33 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 15% cross-reactivity with recombinant human A33 is observed. | |
| Mouse | |
| IgG |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| A33, cell surface A33 antigen, Glycoprotein A33, glycoprotein A33 (transmembrane), GPA33, MGC129986, MGC129987, transmembrane glycoprotein A33 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 10223 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction