missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse A33 Antibody, R&D Systems™

Goat Polyclonal Antibody
£167.00 - £442.00
Specifications
| Antigen | A33/GPA33 |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18798307
![]() |
R&D Systems
AF2756-SP |
25 μg |
£167.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18089851
![]() |
R&D Systems
AF2756 |
100 μg |
£442.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
A33/GPA33 Polyclonal specifically detects A33/GPA33 in Mouse samples. It is validated for Western Blot, Immunohistochemistry.Specifications
| A33/GPA33 | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| Polyclonal | |
| Goat | |
| Mouse | |
| A33, cell surface A33 antigen, Glycoprotein A33, glycoprotein A33 (transmembrane), GPA33, MGC129986, MGC129987, transmembrane glycoprotein A33 | |
| GPA33 | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| LYOPH | |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| RUO | |
| Q9JKA5 | |
| 10223 | |
| Mouse myeloma cell line NS0-derived recombinant mouse A33 Leu22-Ile235 Accession # Q9JKA5 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects mouse A33 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 15% cross-reactivity with recombinant human A33 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title